Merck
  • Selecting the right gate to identify relevant cells for your assay: a study of thioglycollate-elicited peritoneal exudate cells in mice.
Selecting the right gate to identify relevant cells for your assay: a study of thioglycollate-elicited peritoneal exudate cells in mice.
BMC research notes (2017-12-07)
Micely D R Hermida, Rafaela Malta, Marcos D P C de S Santos, Washington L C Dos-Santos
ABSTRACT
In this study, we investigate the diversity and modulation of leukocyte populations represented in the gates defined by size and granularity at different time points of thioglycollate-induced peritonitis in mouse. The inflammatory cells were distributed into four regions (R1-R4) of a data plot graph defined by cell size and granularity. R1 and R2 contained agranular cells that were small in size and predominately included T (CD3+) lymphocytes along with B (B220+) lymphocytes. Macrophages (F4/80+) were the predominant cells found in the R3 region. However, these cells were present in all regions, albeit at a lower frequency in R1 and R2. Granulocytes (Gr1+) were mainly distributed in R3 and R4. The wide distribution of F4/80+ and Gr1+ cells may reflect the recruitment and activation state of the different macrophage and granulocyte populations. Based on these observations, size and granularity may contribute to an initial step in the analysis and sorting of thioglycollate-elicited peritoneal exudate cells. However, the developmental stage and cell activation state may interfere with cell segregation using size and granularity as parameters.