- A cost-effective pH-sensitive release system for water source pH detection.
A cost-effective pH-sensitive release system for water source pH detection.
Chemical communications (Cambridge, England) (2014-09-30)
Zhaoliang Zheng, Xing Huang, Dmitry Shchukin
PMID25262925
ABSTRACT
A facile and cost-effective strategy has been developed to form basic cobalt carbonate nanovalves at the orifice of mesoporous nanocontainers, which facilitate the pH sensitive release of functional cargo for up-scaling towards applications in water source pH detection.