Merck
  • Thermostability detection and optimization of glycoengineered antibodies and antibody-drug conjugates based on differential scanning flouremitry analysis.
Thermostability detection and optimization of glycoengineered antibodies and antibody-drug conjugates based on differential scanning flouremitry analysis.
Bioorganic chemistry (2019-11-26)
Ken Qin, Wei Shi, Lei Zhao, Mingjie Li, Yubo Tang, Faridoon, Bofeng Jiang, Feng Tang, Wei Huang
ABSTRACT
Thermostability of monoclonal antibodies (mAbs) and antibody-drug conjugates (ADCs), as a critical property of biotherapeutics, is important for their physicochemical processes, pharmacodynamics, and pharmacokinetics. Fc glycosylation of mAbs plays a crucial role in antibody functions including thermostability, however, due to the lack of homogeneous glycosylation for comparison, the precise impact of glycoforms on thermostability of mAbs and ADCs remains challenging to elucidate. In this paper, we employed the technique of differential scanning fluorimetry (DSF) to investigate the thermostability of Fc domains, glycoengineered mAbs, and ADCs, carrying well-defined N-glycan structures for comparison. The results revealed that high-mannose-type N-glycans dramatically reduce the Tm value of Fc, compared to complex-type N-glycans. We also found that core-fucose contributes to the thermostability of mAbs, and the unnatural modification on non-reducing end of biantennary N-glycan can compensate the reduced stability of afucosylated mAbs and maintain the advantage of enhanced antibody-dependent cell-mediated cytotoxicity (ADCC). DSF analysis of lysine-linked and glycosite-specific ADCs indicated that thermostability of glycan-linked ADCs is reduced, but it could be improved by using an optimized linkage. This work provides an in-depth analysis on thermostability of mAbs and ADCs with homogeneous glycoforms, and also proposes new strategies for optimizing glycoengineered mAbs and glycosite-specific ADCs using unnatural glycan and stabilized linkage.
Sigma-Aldrich
2-Chloro-1,3-dimethylimidazolinium chloride