Merck
  • Treatment with IL-19 improves locomotor functional recovery after contusion trauma to the spinal cord.
Treatment with IL-19 improves locomotor functional recovery after contusion trauma to the spinal cord.
British journal of pharmacology (2018-03-04)
Jidong Guo, Huadong Wang, Li Li, Yanli Yuan, Xiuxiu Shi, Shuxun Hou
ABSTRACT
IL-19 skews the immune response towards a Th2 type and appears to stimulate angiogenesis. In the current study, we tested if IL-19 treatment could reduce secondary injury and improve functional recovery after contusion spinal cord injury (SCI). Firstly, mice were given a moderate-severe thoracic SCI at the T9-10 level and expression of IL-19 and its receptor was measured in the injured spinal cord. Then SCI mice were treated with mouse recombinant IL-19 and its blocking antibody to investigate the therapeutic effect of IL-19. Protein expression of IL-19 and its receptor IL-20R1 and IL-20R2 was up-regulated in the injured spinal cord of mice. IL-19 treatment promoted the recovery of locomotor function dose-dependently and reduced loss of motor neurons and microglial and glial activation following SCI. Treatment of SCI mice with IL-19 attenuated macrophage accumulation, reduced protein levels of TNF-α and CCL2 and promoted Th2 response and M2 macrophage activation in the injured region. Treatment of SCI mice with IL-19 promoted angiogenesis through up-regulating VEGF in the injured region. Treatment of SCI mice with IL-19 up-regulated HO-1 expression and decreased oxidative stress in the injured region. The beneficial effect of IL-19 was abolished by coadministration of the blocking antibody. Additionally, IL-19 deficiency in mice delayed the recovery of locomotor function following SCI. IL-19 treatment reduced secondary injuries and improved locomotor functional recovery after contusion SCI, through diverse mechanisms including immune cell polarization, angiogenesis and anti-oxidative responses.
Product Description
Buprenorphine, European Pharmacopoeia (EP) Reference Standard