Skip to Content
Merck
α-Tocopherol British Pharmacopoeia (BP) Reference Standard

Alpha Tocopherol

α-Tocopherol British Pharmacopoeia (BP) Reference Standard
Synonym(s):
(±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E
Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique