Sodium dodecyl sulfate solution
Synonym(s):
Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate solution, Sodium n-Dodecyl Sulfate, 20% Solution (w/v)
Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Products
Product Category
Brand
Sustainability
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing