
Hydrogen bromide

Synonym(s):
Hydrobromic acid
Linear Formula:
HBr
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing