Diisobutylaluminum deuteride solution
Synonym(s):
DIBAL-D
Linear Formula:
[(CH3)2CHCH2]2AlD
Molecular Weight:
143.22
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing