Skip to Content
Merck
Titanium(IV) oxide nanowires, diam. × L ~100 nm × 10 μm

Titanium(IV) oxide

Titanium(IV) oxide nanowires, diam. × L ~100 nm × 10 μm
Synonym(s):
Titania, Titanium dioxide, Titanium(IV) oxide
Linear Formula:
TiO2
CAS Number:
Molecular Weight:
79.87
EC Number: