
4,4′-Dibromooctafluorobiphenyl

Linear Formula:
BrC6F4C6F4Br
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing