Skip to Content
Merck
3-Bromo-2-(bromomethyl)propionic acid 97%

3-Bromo-2-(bromomethyl)propionic acid

3-Bromo-2-(bromomethyl)propionic acid 97%
Synonym(s):
β,β′-Dibromoisobutyric acid
Linear Formula:
(BrCH2)2CHCO2H
CAS Number:
Molecular Weight:
245.90

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing