Skip to Content
Merck
1-Boc-3-piperidone 97%

1-Boc-3-piperidone

1-Boc-3-piperidone 97%
Synonym(s):
tert-Butyl 3-oxopiperidine-1-carboxylate
Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing