L-Histidine
Synonym(s):
(S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773
Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Technique