Skip to Content
Merck
Actinomycin D from Streptomyces sp., ≥95% (HPLC)

Actinomycin D

Actinomycin D from Streptomyces sp., ≥95% (HPLC)
Synonym(s):
Actinomycin C1, Actinomycin IV, Dactinomycin
Empirical Formula (Hill Notation):
C62H86N12O16
CAS Number:
Molecular Weight:
1255.42
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Technique
Showing 1-2 of 2