Skip to Content
Merck
p-Xylene anhydrous, ≥99%

p-Xylene

p-Xylene anhydrous, ≥99%
Synonym(s):
1,4-Dimethylbenzene, p-Xylene
Linear Formula:
C6H4-1,4-(CH3)2
CAS Number:
Molecular Weight:
106.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-6 of 6