25717
Untreated Fused Silica Tubing
L × I.D. 3.0 m × 0.25 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115710
material
fused silica tubing
packaging
pkg of 1 ea
df
0.25 μm
technique(s)
gas chromatography (GC): suitable
L × I.D.
3.0 m × 0.25 mm
Looking for similar products? Visit Product Comparison Guide
Other Notes
We offer a variety of chromatography accessories including analytical syringes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service