Skip to Content
Merck
All Photos(1)

Key Documents

NS20Y

8062517, mouse nerve, Neuronal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

NS20Y, 08062517

biological source

mouse nerve

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Choline acetyltransferase

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma

Cell Line Description

The cell line NS20Y is a cholinergic cell line derived from mouse neuroblastoma C1300 (Mouse strain A/Jax).

Application

can be used as a neuronal model

Culture Medium

DMEM + 2 mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service