Skip to Content
NL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
MystiCq
®
microRNA
®
SYBR
®
Green qPCR ReadyMix
™
Products
Brand
Sigma-Aldrich (4)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MIRRM03
MystiCq
®
microRNA
®
SYBR
®
Green qPCR ReadyMix
™
,
Formulation for miRNA RT-qPCR on iQ Bio-Rad platforms
Expand
View Pricing
MIRRM02
MystiCq
®
microRNA
®
SYBR
®
Green qPCR ReadyMix
™
,
with ROX
™
, formulation for miRNA RT-qPCR
Expand
View Pricing
MIRRM00
MystiCq
®
microRNA
®
SYBR
®
Green qPCR ReadyMix
™
,
Formulated for systems that do not require normalization
Expand
View Pricing
MIRRM01
MystiCq
®
microRNA
®
SYBR
®
Green qPCR ReadyMix
™
,
Low ROX
™
formulation for miRNA RT-qPCR
Expand
View Pricing