GF65395031
Tantalum
tube, 100mm, outside diameter 2.0mm, inside diameter 1mm, wall thickness 0.5mm, as drawn, 99.9%
Synonym(s):
Tantalum, TA007100, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.9%
form
tubes
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 653-950-31
resistivity
13.5 μΩ-cm, 20°C
L × wall thickness
100 mm × 0.5 mm
O.D. × I.D.
2.0 mm × 1 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Robert Cohen
American journal of orthopedics (Belle Mead, N.J.), 31(4), 216-217 (2002-05-15)
Conventional materials used for orthopedic reconstruction implants limit the design of new and improved implants. Solid metal is rigid. Porous fixation surfaces have low volumetric porosity available for biologic ingrowth and low frictional characteristics for initial implant stability and have
J Black
Clinical materials, 16(3), 167-173 (1993-12-09)
A detailed literature search was carried out to define the current knowledge about the biological performance of tantalum. The pure metal appears, to a great degree, to be inert both in vivo and in vitro. Both the pure metal and
Jian Lu et al.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 26(2), 244-247 (2012-03-13)
To review the progress in the treatment of bone defect by porous tantalum implant. Recent literature was extensively reviewed and summarized, concerning the treatment method of bone defect by porous tantalum implant. By right of their unique properties, porous tantalum
Juncai Deng et al.
Sheng wu yi xue gong cheng xue za zhi = Journal of biomedical engineering = Shengwu yixue gongchengxue zazhi, 28(2), 419-422 (2011-05-25)
This paper describes the biological characteristics of the metal tantalum (Ts) and the application status of tantalum in artificial joints, bone necrosis, spine, defects of bone and other aspects of bone. The early clinical application results of tantalum in bone
Giulio Maccauro et al.
Recent patents on biotechnology, 3(3), 157-165 (2009-06-24)
Tantalum is obtained from the minerals colombite, tantalite and euxenite. It is greyish silver, heavy and very hard. Tantalum does not react with body fluids and is used to make surgical equipment. Tantalum also does not irritate the body and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service