Demeclocycline HCl
Synonym(s):
Demeclocycline hydrochloride, 7-Chloro-6-demethyltetracycline hydrochloride, Demethylchlorotetracycline hydrochloride, Chlorodemethyltetracycline hydrochloride
Empirical Formula (Hill Notation):
C21H21ClN2O8 · HCl
CAS Number:
Molecular Weight:
501.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique