Skip to Content
Merck
All Photos(1)

Key Documents

V800328

Sigma-Aldrich

Potassium disulfite

AR, ≥97%

Synonym(s):

Potassium metabisulfite, Potassium pyrosulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O5
CAS Number:
Molecular Weight:
222.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
powder

grade

AR

product line

Vetec

Assay

≥97%

form

powder

pH

3.0-4.5 (5% in water)

anion traces

chloride (Cl-): ≤0.005%

cation traces

Fe: ≤0.0005%
heavy metals (as Pb): ≤0.001%

SMILES string

[K+].[K+].[O-]S(=O)S([O-])(=O)=O

InChI

1S/2K.H2O5S2/c;;1-6(2)7(3,4)5/h;;(H,1,2)(H,3,4,5)/q2*+1;/p-2

InChI key

RWPGFSMJFRPDDP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Takahashi et al.
Japanese journal of cancer research : Gann, 77(2), 118-124 (1986-02-01)
Ethanol, potassium metabisulfite, formaldehyde and hydrogen peroxide were tested for tumor-promoting activity in a two-stage stomach carcinogenesis experiment. Male outbred Wistar rats were given N-methyl-N'-nitro-N-nitrosoguanidine (MNNG) in the drinking water (100 mg/liter) and a diet supplemented with 10% sodium chloride
W H Yang et al.
CMAJ : Canadian Medical Association journal = journal de l'Association medicale canadienne, 133(9), 865-867 (1985-11-01)
Sulfites are widely used as preservatives in the food and pharmaceutical industries. In the United States more than 250 cases of sulfite-related adverse reactions, including anaphylactic shock, asthmatic attacks, urticaria and angioedema, nausea, abdominal pain and diarrhea, seizures and death
Luca Settanni et al.
Journal of bioscience and bioengineering, 114(6), 606-614 (2012-08-11)
In this work, the yeast ecology associated with the spontaneous fermentation of Grillo cultivar grapes from 10 vineyards was analyzed from grape harvest till complete consumption of must sugars. The microbiological investigation started with the plate count onto two culture
Ayse Yavuz-Kocaman et al.
Environmental and molecular mutagenesis, 49(4), 276-282 (2008-03-14)
Potassium metabisulfite (PMB) is used as an antimicrobial substance in many kinds of foods. In the present study, the effects of PMB on chromosome aberrations (CAs), sister chromatid exchanges (SCEs), and micronucleus (MN) formation in human lymphocytes and as well
Taylor C Wallace et al.
Journal of food science, 76(7), C1010-C1017 (2012-03-16)
Cacao (Theobroma cacao L.) is rich in procyanidins, a large portion of which degrades during the natural fermentation process of producing cocoa powder. Recent advances in technology have enabled scientists to produce unfermented cocoa powder, preserving the original profile of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service