V800214
Hydroxylamine hydrochloride
AR, ≥99%
Synonym(s):
Hydroxylammonium chloride
About This Item
Recommended Products
grade
AR
vapor pressure
0.001 hPa ( 50 °C)
product line
Vetec™
Assay
≥99%
form
crystals
pH
2.5-3.5 (20 °C, 50 g/L)
mp
155-157 °C (dec.) (lit.)
density
1.67 g/mL at 25 °C (lit.)
anion traces
sulfate (SO42-): ≤0.002%
cation traces
Fe: ≤0.0005%
NH4+: ≤0.10%
heavy metals (as Pb): ≤0.0005%
SMILES string
Cl.NO
InChI
1S/ClH.H3NO/c;1-2/h1H;2H,1H2
InChI key
WTDHULULXKLSOZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral
Target Organs
spleen
Storage Class Code
4.1A - Other explosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service