Skip to Content
Merck
All Photos(1)

Key Documents

V800179

Sigma-Aldrich

1,2-Dichloroethane

AR, ≥99.5%

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
AR
bp:
83 °C (lit.)
vapor pressure:
87 mmHg ( 25 °C)

grade

AR

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

dilution

(for analytical testing)

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Perreault et al.
Chemical Society reviews, 44(16), 5861-5896 (2015-03-27)
Graphene-based materials are gaining heightened attention as novel materials for environmental applications. The unique physicochemical properties of graphene, notably its exceptionally high surface area, electron mobility, thermal conductivity, and mechanical strength, can lead to novel or improved technologies to address
Rongke Gao et al.
Biosensors & bioelectronics, 72, 230-236 (2015-05-20)
We report the application of a fully automated surface-enhanced Raman scattering (SERS)-based solenoid-embedded microfluidic device to the quantitative and sensitive detection of anthrax biomarker poly-γ-D-glutamic acid (PGA) in solution. Analysis is based on the competitive reaction between PGA and PGA-conjugated
Delphine Schaming et al.
Physical chemistry chemical physics : PCCP, 13(39), 17704-17711 (2011-09-13)
Photocurrent responses associated with the interfacial quenching of the photo-excited water-soluble zinc meso-tetra(4-carboxyphenyl)porphyrin (ZnTPPC) by ferrocene have been studied at a water|1,2-dichloroethane interface in the absence and in the presence of adsorbed gold nanoparticles. Upon addition of methanol, a mirror-like
Hitokazu Esaki et al.
The Journal of allergy and clinical immunology, 135(1), 153-163 (2015-01-09)
The molecular signature of atopic dermatitis (AD) lesions is associated with TH2 and TH22 activation and epidermal alterations. However, the epidermal and dermal AD transcriptomes and their respective contributions to abnormalities in respective immune and barrier phenotypes are unknown. We
Leila Mahmoudi et al.
Dalton transactions (Cambridge, England : 2003), 40(34), 8695-8700 (2011-08-02)
Manganese(III)-meso-tetraphenylporphyrin [Mn(TPP)] and manganese(III)-meso-tetrakis(pentafluorophenyl)porphyrin [Mn(TPFPP)] catalyse the epoxidation of cyclooctene by IO(4)(-) in the presence of excess imidazoles, in both dry CH(2)Cl(2) and CH(2)Cl(2) saturated with H(2)O. The reaction rates of the electron deficient Mn(TPFPP) are a factor 24 less

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service