Skip to Content
Merck
All Photos(1)

Key Documents

V800153

Sigma-Aldrich

Diethanolamine

LR, ≥98%

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
Assay:
≥98%
bp:
217 °C/150 mmHg (lit.)
vapor pressure:
<0.98 atm ( 100 °C)

grade

LR

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

product line

Vetec

Assay

≥98%

form

viscous liquid

autoignition temp.

689 °F

expl. lim.

10.6 %

refractive index

n20/D 1.477 (lit.)

pH

11.0-12 (25 °C, 105 g/L)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-Ichi Kusumoto et al.
Journal of biomedical materials research. Part A, 91(1), 1-10 (2008-08-07)
Understanding of lipidic materials used for gene delivery system is essential for the effective design and development of potential applications in basic and therapeutic research. This study aimed to evaluate the biological activity of totally synthesized ditetradecylacetyldiethanolaminetrimethylammonium (TMA-C2-DEA-C14) as gene
Shiguo Sun et al.
Dalton transactions (Cambridge, England : 2003), (38)(38), 7969-7974 (2009-09-23)
A series of ruthenium(II) tris-bipyridyl complexes covalently linked with different amine reductants such as tripropylamine (TPrA), ethanolamine and diethanolamine for an electrochemiluminescence (ECL) system have been synthesized. Their ECL property at different working electrodes has been studied with and without
William Conway et al.
Environmental science & technology, 46(13), 7422-7429 (2012-05-25)
The kinetics of the fast reversible carbamate formation reaction of CO(2)(aq) with a series of substituted cyclic secondary amines as well as the noncyclic secondary amine diethanolamine (DEA) has been investigated using the stopped-flow spectrophotometric technique at 25.0 °C. The
Siou-Yin Chen et al.
Inorganic chemistry, 51(8), 4448-4457 (2012-04-05)
The synthesis, X-ray crystallography, magnetic properties, and high-field electron paramagnetic resonance (HFEPR) of a new heptanuclear manganese complex [Mn(7)(heamp)(6)](ClO(4))(2)·4CH(2)Cl(2)·H(2)O (complex 2), in which heampH(3) is 2-[N,N-di(2-hydroxyethyl)aminomethyl]phenol (compound 1), is reported. Complex 2 has a hexagonal, disk-shaped topology and contains six
L Jiménez et al.
Bioresource technology, 100(3), 1478-1481 (2008-09-24)
The AOpAZRP bleaching sequence (A is an acid treatment, Op an oxygen and peroxide stage, Z an ozone stage, R a reductive treatment and P a peroxide stage) have been applied to oil palm empty fruit bunches (EFB) soda-anthraquinone and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service