Skip to Content
Merck
All Photos(2)

Key Documents

V800095

Sigma-Aldrich

Cadmium(II) acetate

LR, ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cd(OCOCH3)2
CAS Number:
Molecular Weight:
230.50
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥98%

form

crystalline powder

mp

255-273 °C

SMILES string

CC(=O)O[Cd]OC(C)=O

InChI

1S/2C2H4O2.Cd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

LHQLJMJLROMYRN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - STOT RE 1 Oral

Target Organs

Kidney,Bone

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Saravanan et al.
Journal of colloid and interface science, 452, 126-133 (2015-05-04)
A ternary ZnO/Ag/CdO nanocomposite was synthesized using thermal decomposition method. The resulting nanocomposite was characterized by X-ray diffraction, field emission scanning electron microscopy, transmission electron microscopy, UV-Vis spectroscopy, and X-ray photoelectron spectroscopy. The ZnO/Ag/CdO nanocomposite exhibited enhanced photocatalytic activity under
J I Chao et al.
Mutation research, 498(1-2), 7-18 (2001-10-24)
Previously, we have demonstrated that cadmium acetate significantly induces hprt mutation frequency in Chinese hamster ovary (CHO)-K1 and that 3-amino-1,2,4-triazole (3AT), a catalase inhibitor, potentiates the mutagenicity of cadmium [Chem. Res. Toxicol. 9 (1996) 1360-1367]. In this study, we investigate
L M Cavaco et al.
Antimicrobial agents and chemotherapy, 54(9), 3605-3608 (2010-06-30)
We recently reported a phenotypic association between reduced susceptibility to zinc and methicillin resistance in Staphylococcus aureus CC398 isolates from Danish swine (F. M. Aarestrup, L. M. Cavaco, and H. Hasman, Vet. Microbiol. 142:455-457, 2009). The aim of this study
M Lag et al.
Cell biology and toxicology, 18(1), 29-42 (2002-05-07)
The lung is a target organ for cadmium (Cd) toxicity. Apoptosis induced by cadmium acetate (CdAc) was studied in alveolar type 2 cells and Clara cells isolated from rat lung. Relatively low concentrations of CdAc (1-10 micromol/L) induced apoptosis after
Albert L Juhasz et al.
Environmental science & technology, 44(13), 5240-5247 (2010-06-10)
In this study, cadmium (Cd) relative bioavailability in contaminated (n = 5) and spiked (n = 2) soils was assessed using an in vivo mouse model following administration of feed containing soil or Cd acetate (reference material) over a 15

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service