All Photos(1)
About This Item
Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
powder
Recommended Products
grade
LR
product line
Vetec™
Assay
≥98%
form
powder
pH
7
SMILES string
[Ba++].[O-]S([O-])(=O)=O
InChI
1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
InChI key
TZCXTZWJZNENPQ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mahavir Singh et al.
Canadian journal of physiology and pharmacology, 99(1), 56-63 (2020-08-18)
Epigenetic DNA methylation (1-carbon metabolism) is crucial for gene imprinting/off-printing that ensures epigenetic memory but also generates a copious amount of homocysteine (Hcy), unequivocally. That is why during pregnancy, expectant mothers are recommended "folic acid" preemptively to avoid birth defects
George X Huang et al.
Tissue engineering. Part A, 21(15-16), 2228-2240 (2015-04-30)
Polymeric scaffolds, which release growth factors in a temporally controlled manner, have successfully directed the differentiation of stem cells into monolithic tissues of a single lineage. However, engineering precise boundaries in multilineage functional tissues, such as the juxtaposed cartilaginous and
Koji Murono et al.
Diseases of the colon and rectum, 57(6), 715-724 (2014-05-09)
Preoperative chemoradiotherapy has been widely used for the prevention of local recurrence of locally advanced rectal cancer, and the effect of chemoradiotherapy is known to be associated with overall survival. We aimed to evaluate the association of the pathologic response
Dewey C Scheid et al.
Journal of the American Board of Family Medicine : JABFM, 26(5), 498-507 (2013-09-06)
The purpose of this study was to describe colorectal cancer screening (CRCS) practices across a variety of primary care clinics and identify the methods used by primary care physicians (PCPs) with higher rates of CRCS ("exemplars"). Physician questionnaires, structured interviews
Joon-Sung Kim et al.
American journal of physical medicine & rehabilitation, 92(10), 912-919 (2013-06-07)
The aim of this study was to report the characteristics of dysphagia in children with cerebral palsy (CP), related to gross motor function. Videofluoroscopic swallow study was performed in 29 children with CP, according to the manual of Logemann. Five
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service