Skip to Content
Merck
All Photos(1)

Key Documents

V800062

Sigma-Aldrich

Barium acetate

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ba
CAS Number:
Molecular Weight:
255.42
Beilstein:
3693411
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

7-8 (25 °C, 50 g/L)

SMILES string

CC(=O)O[Ba]OC(C)=O

InChI

1S/2C2H4O2.Ba/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

ITHZDDVSAWDQPZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edwige Van der Heiden et al.
Journal of molecular microbiology and biotechnology, 25(2-3), 106-119 (2015-07-15)
We report the first enzymatic synthesis of D-tagatose-1-phosphate (Tag-1P) by the multicomponent phosphoenolpyruvate:sugar phosphotransferase system (PEP-PTS) present in tagatose-grown cells of Klebsiella pneumoniae. Physicochemical characterization by (31)P and (1)H nuclear magnetic resonance spectroscopy reveals that, in solution, this derivative is
Li-Chan Yang et al.
Carbohydrate polymers, 124, 150-156 (2015-04-04)
Rice hulls (Oryza sativa) are high in carbohydrate content and have been utilized as dietary fiber. The immunomodulatory bioactivity of rice hull polysaccharides (RHPS) has rarely been reported. This study demonstrated the structural characteristics and immunomodulating of RHPS. The RHPS
Li-Chan Yang et al.
Carbohydrate polymers, 111, 856-863 (2014-07-20)
Dendrobium tosaense is a medicinal Dendrobium species widely used in traditional medicine. This study demonstrated some structural characterizations and immunomodulatory activity of the water-soluble polysaccharides derived from the stem of D. tosaense (DTP). DTP was fractioned using DEAE-650 M anion-exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service