Skip to Content
Merck
All Photos(1)

Key Documents

V800026

Sigma-Aldrich

Aluminum potassium sulfate dodecahydrate

LR, ≥99%

Synonym(s):

Alum, Potassium alum, Potassium aluminum sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlK(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
474.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

3.3 (94.88 g/L)

mp

92 °C (lit.)

density

1.757 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saya Moriyama et al.
The Journal of experimental medicine, 211(7), 1297-1305 (2014-06-11)
Follicular helper T (Tfh) cells access the B cell follicle to promote antibody responses and are particularly important for germinal center (GC) reactions. However, the molecular mechanisms of how Tfh cells are physically associated with GCs are incompletely understood. We
Gareth R Williams et al.
The Journal of experimental medicine, 211(6), 1019-1025 (2014-05-07)
There is currently no paradigm in immunology that enables an accurate prediction of how the immune system will respond to any given agent. Here we show that the immunological responses induced by members of a broad class of inorganic crystalline
Yan-Feng Li et al.
Nature communications, 5, 4273-4273 (2014-07-01)
Germline or B-cell-specific loss of Ptpn6 gene encoding the Shp1 protein tyrosine phosphatase leads to skewed B lymphopoiesis and systemic autoimmunity. Here, to study its role in B-cell terminal differentiation, we generated Ptpn6(f/f)Aicda(Cre/+) mice with Shp1 ablated only in activated
Bettina Grötsch et al.
The Journal of experimental medicine, 211(11), 2199-2212 (2014-10-08)
The cornerstone of humoral immunity is the differentiation of B cells into antibody-secreting plasma cells. This process is tightly controlled by a regulatory gene network centered on the transcriptional repressor B lymphocyte-induced maturation protein 1 (Blimp1). Proliferation of activated B
K Yadava et al.
Allergy, 69(10), 1333-1342 (2014-06-26)
Thymic stromal lymphopoietin (TSLP) is a cytokine primarily produced by epithelial cells, which has been shown to be a potent inducer of T-helper 2 (Th2)-type responses. However, TSLP has pleiotropic effects upon immune cells, and although extensively studied in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service