Skip to Content
Merck
All Photos(2)

Key Documents

V800009

Sigma-Aldrich

1-Naphthol

LR, ≥99%

Synonym(s):

α-Naphthol, 1-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein:
1817321
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

LR

vapor density

4.5 (120 °C, vs air)

vapor pressure

1 mmHg ( 94 °C)

product line

Vetec

Assay

≥99%

form

crystals

autoignition temp.

1007 °F

expl. lim.

5 %

bp

278-280 °C (lit.)

mp

94-96 °C (lit.)

SMILES string

Oc1cccc2ccccc12

InChI

1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H

InChI key

KJCVRFUGPWSIIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 2 Oral - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ondrej Zitka et al.
Electrophoresis, 32(22), 3207-3220 (2011-10-21)
Guanosine derivatives are important for diagnosis of oxidative DNA damage including 8-hydroxy-2'-deoxyguanosine (8-OHdG) as one of the most abundant products of DNA oxidation. This compound is commonly determined in urine, which makes 8-OHdG a good non-invasive marker of oxidation stress.
Anika Kasprick et al.
European journal of immunology, 45(5), 1462-1470 (2015-02-14)
The role of mast cells (MCs) in autoimmunity is the matter of an intensive scientific debate. Based on observations in different MC-deficient mouse strains, MCs are considered as fundamental players in autoimmune diseases. However, most recent data suggest that the
Shitong Yang et al.
The Science of the total environment, 420, 214-221 (2012-02-15)
The increasing exploitation of multi-walled carbon nanotubes (MWCNTs) into many industrial processes has raised considerable concerns as they are likely to be released into the environment. The interactions of the pollutants in water with discharged MWNCTs will further influence the
Xiaoying Guo et al.
Environmental science & technology, 46(13), 7252-7259 (2012-06-09)
The sorption behavior of four hydrophobic organic contaminants (HOCs) (i.e., phenanthrene, naphthalene, lindane, and 1-naphthol) by three types of polymers namely polyethylene (PE), polystyrene (PS), and polyphenyleneoxide (PPO) was examined in this work. The organic carbon content-normalized sorption coefficients (K(oc))
Hongbo Zhang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(8), 1286-1293 (2012-01-24)
In vitro glucuronidation assays of diclofenac and indomethacin at pH 7.4 are biased by the instability of the glucuronides due to acyl migration. The extent of this acyl migration may be reduced significantly by performing the glucuronidation reaction at pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service