material
polyester film
sterility
non-sterile
packaging
pack of 100 ea
manufacturer/tradename
Excel Scientific 100-THIN-PLT
technique(s)
ELISA: suitable
L
141 mm
W × L
7.94 cm × 14.1 cm
W
79.4 mm
General description
Semi-pierceable, and leaves minimal residue. Polyester films just 25 μm thick, clearer and marginally pierceable with single-channel pipettes for sample recovery. Dimensions are 54.0 mm x 141.0 mm.
Legal Information
ThinSeal is a trademark of Excel Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)