GF80427843
Lead
foil, not light tested, 100x100mm, thickness 0.03mm, 99.99+%
Synonym(s):
Lead, PB000170
About This Item
Assay
99.99%
form
foil
manufacturer/tradename
Goodfellow 804-278-43
resistivity
20.65 μΩ-cm
size × thickness
100 x 100 mm × 0.03 mm
bp
1740 °C (lit.)
mp
327.4 °C (lit.)
SMILES string
[Pb]
InChI
1S/Pb
InChI key
WABPQHHGFIMREM-UHFFFAOYSA-N
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral
Target Organs
Central nervous system,Blood,Immune system,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service