509833
SGE Microvolume Syringe for Rheodyne® and Valco® Valves, Fixed Needle
500F-LC, volume 500 μL, needle size 22 ga, needle L × O.D. 2 in. × 0.7112 mm (0.028 in.)
About This Item
UNSPSC Code:
41122001
material
blunt tip stainless steel needle (point style 3)
needle size
22 ga
feature
Fixed needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 007300
needle L × O.D.
2 in. × 0.7112 mm (0.028 in.)
volume
500 μL
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)