Documentos clave
Z640069
Eppendorf® epT.I.P.S. standard
volume range 0.1-10 μL
Sinónimos:
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
gray
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (2 bags of 500 tips)
manufacturer/tradename
Eppendorf® 0030 000.811
Eppendorf® 022492004
L
34 mm
maximum volume
10 μL
volume range
0.1-10 μL
General description
Bulk packed in re-sealable bags.
200 μL, 300 μL and 1000 μL tips are color coded, yellow, orange and blue respectively.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG