Documentos clave
Z334138
BRAND® cryovial racks, PP
white
About This Item
UNSPSC Code:
41122806
NACRES:
NB.12
material
polypropylene
white
feature
positions 6 × 14
manufacturer/tradename
BRAND 4341000
BRAND
technique(s)
cryopreservation: suitable
L × W × H
265 mm × 126 mm × 38 mm
diam.
13 mm
General description
Racks for cryo vials are stackable, with alphanumerical positions. Racks are made of PP (filled PP, density 1.2 g/cm3), will not float in waterbath. Operating temperature: -20 to 90 °C. Autoclavable at 121 °C. Sturdy construction racks are supplied in two-pieces for convenient and permanent assembly. Available in four different colors.
Other Notes
BR4341000 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)