Documentos clave
Z331422
Lenz® round bottom flasks
capacity 100 mL, joint: ST/NS 14/23
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass 3.3
round bottom flask
single-neck
joint
joint: ST/NS14/23
agency
according to DIN 12 348
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Lenz 3.0014.37
capacity
100 mL
Legal Information
Lenz is a registered trademark of Lenz GmbH
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)