Merck
H9759
Sigma-Aldrich
Derivatized Silica Gel
40-63 μm mean particle size, average diameter 60 Å
About This Item
Número de CAS:
MDL number:
UNSPSC Code:
41115700
form
powder
Quality Level
extent of labeling
~22% carbon loading
surface area
500 m2/g
mean particle size
40-63 μm
pore size
60 Å average diameter
Preparation Note
Prepared by the reaction of silica gel with n-octadecyltrichlorosilane. After hydrolysis of the unreacted chloro groups, hydroxyl groups are blocked with trimethylsilyl groups.
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 1