Documentos clave
GE28-9666-46
Sigma-Aldrich
Column tube assembly
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
technique(s)
LPLC: suitable
Features and Benefits
Column tube consist of both the inner glass tube, the protection tube and the tube holder
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)