Documentos clave
EP5825711009
Eppendorf® Rotor Accessories
plate adapter, capacity, 1 x 96 well PCR plate, pack of 2
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
capacity 1 x 96 well PCR plate
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5825711009
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with MTP/Flex bucket
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)