BAF188510017
Bel-Art® Freezer Storage Box
no. of holes, 100, black polypropylene, pack of 5
Sinónimos:
cryopreservation storage rack, cryovial storage, indexed tube rack, microtube storage
About This Item
Productos recomendados
material
black polypropylene
feature
for tubes 0.5, 1.5, and 2.0 mL
no. of holes 100
packaging
pack of 5
manufacturer/tradename
Bel-Art F18851-0017
General description
Features and Benefits
- Alphanumeric Indexing
- Translucent lid for easy access and viewing; matte panels can be used for labeling (100-place boxes have hinged lid, 81-place boxes have lift off lid)
- Withstand -80°C to +121°C (-112° to +249°F)
- Stackable with molded-in ′no slide′ features
- Assorted Colors with translucent lids (natural, blue, green, orange and purple)
Legal Information
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Global Trade Item Number
Número de referencia del producto (SKU) | GTIN |
---|---|
BAF188510017-5EA | 4061832258997 |