Documentos clave
AXYP96450RBK
Round bottom assay plate
size 96 wells, round bottom black polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs
Sinónimos:
96 multiwell plate
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material:
round bottom black polypropylene wells
maximum volume:
500 μL
size:
96 wells
working volume:
450 μL
material
round bottom black polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc P-96-450R-BK
Corning
packaging
pkg of 10x5plates/cs
maximum volume
500 μL
size
96 wells
working volume
450 μL
binding type
non-treated surface
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)