ZWACID012
ROCare
Acid cleaning chemical removes scaling from RO membranes
Synonym(s):
ROCare A
About This Item
Recommended Products
Quality Level
feature
removes scaling from RO membranes
packaging
pkg of 12 packs
compatibility
for use with Elix® Advantage
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The acid cleaner is delivered in a pouch and the empty pouch is removed after use. It is to be used periodically when prompted by the system, or when the RO % rejection drops more than 5%, or when the RO permeate flow rate drops more than 10% due to mineral scaling.
The ROCare A pouch cleans RO membranes supporting Milli-Q® HR/HX/CLX, Milli-Q® IX 7003/05/10/15, and Elix® Advantage water purification systems.
Application
Features and Benefits
- Anti-scaling agent
- pH 2 in an aqueous solution
- Contains 12 packages
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service