Skip to Content
Merck
All Photos(2)

Key Documents

O2256

Sigma-Aldrich

Oxolamine citrate salt

Synonym(s):

5-(2-[Diethylamino]ethyl)-3-phenyl-1,2,4-oxadiazole citrate salt, NSC 100298

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19N3O · C6H8O7
CAS Number:
Molecular Weight:
437.44
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCN(CC)CCc1nc(no1)-c2ccccc2

InChI

1S/C14H19N3O.C6H8O7/c1-3-17(4-2)11-10-13-15-14(16-18-13)12-8-6-5-7-9-12;7-3(8)1-6(13,5(11)12)2-4(9)10/h5-9H,3-4,10-11H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

RBZIGQJSMCOHSS-UHFFFAOYSA-N

Application

Molecule involved in pharmacological studies including:
  • Antiinflammatory agents
  • Drugs that shift energy metabolism from mitochondrial respiration to glycolysis
  • Effect on warfarin anticoagulant properties

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Therapeutic action of oxolamine in acute tracheo-bronchitis].
G Gunella et al.
La Clinica terapeutica, 100(5), 485-492 (1982-03-15)
L Kirilmaz et al.
Journal of microencapsulation, 9(2), 167-172 (1992-04-01)
One of the principal uses of microencapsulation for pharmaceuticals has been the preparation of sustained-release dosage forms which have been usually presented in the form of a suspension or gel. However, a non-disintegrating tablet would be a better formulation to
Xuan Zhu et al.
Biopharmaceutics & drug disposition, 28(3), 125-133 (2007-02-14)
The possible reason for the significantly greater AUC of oral warfarin with oral oxolamine in male Sprague-Dawley rats was evaluated. After oral administration of warfarin at a dose of 2 mg/kg to male rats with oxolamine at doses of 10
R Cerqua et al.
Minerva medica, 72(7), 355-362 (1981-02-28)
A single-blind, between-patients trial was run to compare the fluidifying, expectorant, antitussive, antipyretic and anti-inflammatory activity, and the local and systemic tolerance of guacetisal, an active principle obtained by the esterification of acetylsalicylic acid and guaiacol, and oxolamine phosphate, in
J McEwen et al.
The Medical journal of Australia, 150(8), 449-450 (1989-04-17)
Twenty reports in Australia, Belgium and The Netherlands have implicated oxolamine-citrate cough-mixtures as a cause of hallucinations in children of less than 10 years of age. Fever is not thought to be a likely alternative explanation. Similar reactions in older

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service