Skip to Content
Merck
All Photos(2)

Key Documents

GF04751265

Silver

foil, 10mm disks, thickness 1.0mm, as rolled, 99.95+%

Synonym(s):

Silver, AG000475

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12141740
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 047-512-65

resistivity

1.59 μΩ-cm, 20°C

diam. × thickness

10 mm × 1.0 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

density

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IX. Note on Silver Foil in Surgery.
J S Lewis
Annals of surgery, 50(4), 793-796 (1909-10-01)
Clinical evaluation of C-Film, a vaginal contraceptive.
O Frankman et al.
The Journal of international medical research, 3(4), 292-296 (1975-01-01)
H H Wiebe et al.
Plant physiology, 59(2), 256-258 (1977-02-01)
Water potential was monitored at nine locations along single maize (Zea mays L.) leaf blades with aluminum block in situ thermocouple hygrometers. Water potential showed a continuous decrease toward the tip, with a 2- to 4-bar difference between leaf base
A Rupérez et al.
Talanta, 44(2), 213-220 (1997-02-01)
Surface-enhanced Raman scattering (SERS) spectrometry of the diuretic drug triamterene is discussed. The SERS-active substrate used is a silver foil etched with nitric acid. The influence of solvent and sample doping method on sensivity, intercept and shape of the calibration
M Dubiel et al.
Journal of synchrotron radiation, 8(Pt 2), 539-541 (2001-08-22)
The thermal expansion behaviour of silver fcc has been described by an anharmonic Einstein model using EXAFS data in the temperature range between 10 and 300 K. The linear expansion coefficient of a bulk silver foil agrees well with X-ray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service