Key Documents
BAF368210110
SP Bel-Art® Sterileware Triangular Scraper
high-density polyethylene, blade L × width 8.875 in. (225 mm) × 11 cm (0.75 in.), sterile
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
high-density polyethylene
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Bel-Art F36821-0110
blade L × width
8.875 in. (225 mm) × 11 cm (0.75 in.)
General description
Sterile, plastic scrapers are perfect for cutting, cleaning and general scraping. Ideal for use in the manufacture and quality assurance of creams, ointments and pastes.
Features and Benefits
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) scrapers heat sealed in individual polyethylene bags
  • Gamma irradiated sterile (SAL 10^-6)
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)