Skip to Content
Merck
All Photos(1)

Key Documents

363197

Sigma-Aldrich

3-Hydroxy-4-methyl-2(3H)-thiazolethione

97%

Synonym(s):

2-Mercapto-4-methylthiazole 3-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5NOS2
CAS Number:
Molecular Weight:
147.22
Beilstein:
1210145
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

92-94 °C (lit.)

solubility

methanol: soluble 25 mg/mL, clear, brownish-yellow to brown

functional group

thioether

SMILES string

CC1=CSC(=S)N1O

InChI

1S/C4H5NOS2/c1-3-2-8-4(7)5(3)6/h2,6H,1H3

InChI key

LOKDIDVKVVVVHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Hydroxy-4-methyl-2(3H)-thiazolethione (HMTT) is a cyclic thiohydroxamic acid. It forms divalent complexes with the first-row d-block elements Co-Zn. Hydrated form of HMTT exhibits enhanced thermal stability and solubility compared with the anhydrous form. HMTT possess a chelating unit similar to pyrithione. Solid state chemistry of HMTT and its complexes with d-block elements has been reported.

Application

3-Hydroxy-4-methyl-2(3H)-thiazolethione may be employed as fungicide and alkoxy-radical precursors in the synthetic procedures. It may be useful in the Barton reaction for the generation of carbon radicals through decarboxylative rearrangement of the corresponding esters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-state study of cyclic thiohydroxamic acids: 1-hydroxy-2 (1H)-pyridinethione and 3-hydroxy-4-methyl-2 (3H)-thiazolethione.
Bond AD and Jones W.
Journal of Physical Organic Chemistry, 13(7), 395-404 (2000)
Divalent complexes of 3-hydroxy-4-methyl-2 (3H)-thiazolethione with Co-Zn: synthesis, X-ray crystal structures and the structure-directing influence of C-HS interactions.
Bond AD and Jones W.
J. Chem. Soc., Dalton Trans., 20, 3045-3051 (2001)
Synthesis and X-ray crystal structures of bis (3-hydroxy-4-methyl-2 (3H)-thiazolethiolato-S 2, O) bis (dimethylsulfoxide-O) M, M= cobalt (II) and nickel (II).
Bond AD and Jones W.
Transition Met. Chem. (London), 27(4), 407-410 (2002)
Journal of the Chemical Society. Perkin Transactions 1, 39-39 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service