SML0324
JW67
≥98% (HPLC)
Synonym(s):
Trispiro[3H-indole-3,2′-[1,3]dioxane-5′,5′′-[1,3]dioxane-2′′,3′′′-[3H]indole]-2,2′′′(1H,1′′′H)-dione (9CI)
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: >5 mg/mL
storage temp.
2-8°C
SMILES string
O=C1Nc2ccccc2C13OCC4(CO3)COC5(OC4)C(=O)Nc6ccccc56
InChI
1S/C21H18N2O6/c24-17-20(13-5-1-3-7-15(13)22-17)26-9-19(10-27-20)11-28-21(29-12-19)14-6-2-4-8-16(14)23-18(21)25/h1-8H,9-12H2,(H,22,24)(H,23,25)
InChI key
BTXRSHKJNDFHGA-UHFFFAOYSA-N
Biochem/physiol Actions
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service