Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100477

Sigma-Aldrich

Anti-CRABP2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CRABP-II, Anti-Cellular retinoic acid binding protein 2, Anti-RBP6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
guinea pig, human
technique(s):
western blot: suitable
citations:
1

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

16 kDa

species reactivity

guinea pig, human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRABP2(1382)

Immunogen

Synthetic peptide directed towards the middle region of human CRABP2

Application

Anti-CRABP2 antibody produced in rabbit is suitable for western blot analysis.

Biochem/physiol Actions

A number of specific carrier proteins for members of the vitamin A family have been discovered. Cellular retinoic acid binding proteins (CRABP) are low molecular weight proteins whose precise function remains unknown. The inducibility of the CRABP2 gene suggests that this isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. It has been postulated that the CRABP2 gene is transcriptionally regulated by a newly synthesized regulatory protein.A number of specific carrier proteins for members of the vitamin A family have been discovered. Cellular retinoic acid binding proteins (CRABP) are low molecular weight proteins whose precise function remains unknown. The inducibility of the CRABP2 gene suggests that this isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. It has been postulated that the CRABP2 gene is transcriptionally regulated by a newly synthesized regulatory protein. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

Sequence

Synthetic peptide located within the following region: FEEQTVDGRPCKSLVKWESENKMVCEQKLLKGEGPKTSWTRELTNDGELI

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benito Campos et al.
International journal of cancer, 131(8), 1963-1968 (2012-01-26)
Impairment of endogenous differentiation pathways like retinoic acid (RA) signaling seems to be a central pathogenetic event in astrocytic gliomas. Among others, expression of the differentiation-promoting RA chaperon protein cellular retinoic acid binding protein 2 (CRABP2) is extenuated in high-grade
Avijit Majumdar et al.
The Journal of biological chemistry, 286(49), 42749-42757 (2011-10-15)
Cellular retinoic acid-binding protein II (CRABP-II) undergoes nuclear translocation upon binding of retinoic acid (RA). In the nucleus, CRABP-II directly binds to the nuclear receptor RAR to form a complex through which RA is "channeled" from the binding protein to
Blendi Ura et al.
Oncology letters, 13(5), 3912-3920 (2017-05-20)
Uterine leiomyoma is the most common female reproductive tract benign tumor. Little is known about protein composition and changes in the leiomyoma interstitial fluid (IF). The present study focused on changes in protein abundance in the IF of leiomyoma. Leiomyoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service