Skip to Content
Merck
All Photos(1)

Documents

MIRAP01424

Sigma-Aldrich

MystiCq® microRNA qPCR Assay Primer

mmu-miR-717

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106308
NACRES:
NA.55

form

liquid

concentration

10 mg/mL

mature sequence

CUCAGACAGAGAUACCUUCUCU

mp

~76.5 °C

Sanger mature/minor accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... MIR717(751531)

General description

The MystiCq® microRNA qPCR Assay Primer is an integral part of the MystiCq microRNA qPCR Assay System. It has been designed to target specific microRNAs with the MystiCq Universal PCR primer and the MystiCq® microRNA SYBR® Green qPCR ReadyMix on cDNA templates generated by the MystiCq microRNA cDNA Synthesis Mix kit.

Features and Benefits

  • Proprietary design features of assay primers allow for specificity towards mature microRNAs vs. precursor-microRNAs
  • Extensive design and testing of oligo-dT adapter primer to incorporate complementary Universal PCR Primer sequence
  • No self-complementarity or primer dimer artifacts with the Universal PCR Primer
  • Optimized primer Tm designed to match the Universal PCR Primer
  • Universal cycling conditions ensures robust amplification for all assays in profiling experiments
  • Optimized PCR product Tm (75-78° C)

Legal Information

MystiCq is a registered trademark of QIAGEN Beverly Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
SYBR is a registered trademark of Life Technologies

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Method for purification, reverse transcription and quantitative PCR for MicroRNAs using Mysticq reagents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service