Recommended Products
Assay
≥98.0% (HPLC)
optical activity
[α]/D +23.0±2°, 24 hr, c = 0.5% in H2O
storage temp.
room temp
SMILES string
OC[C@@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4-,5-,6?/m0/s1
InChI key
WQZGKKKJIJFFOK-QRXFDPRISA-N
Application
L-(+)-Gulose, a rare aldohexose sugar C-3 epimer of galactose, is used in the development of various drugs and to glycosylate alginates. L-(+)-Gulose is a substrate of GDP-mannose 3′,5′-epimerase in plants and may be a precursor of L-ascorbic acid in plants.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Carbohydrate research, 344(11), 1285-1288 (2009-06-09)
An efficient and stereoselective synthesis of D,L-gulose was described. The key step of the synthetic route is represented by a multicomponent enyne cross metathesis-hetero Diels-Alder reaction which allows the formation of the pyran ring from cheap and commercially available substrates
Chemistry (Weinheim an der Bergstrasse, Germany), 14(30), 9400-9411 (2008-09-05)
The glycosylation properties of gulopyranosides have been mapped out, and it is shown that gulose has an intrinsic preference for the formation of 1,2-cis-glycosidic bonds. It is postulated that this glycosylation behaviour originates from nucleophilic attack at the oxacarbenium ion
The Journal of organic chemistry, 77(17), 7401-7410 (2012-08-03)
A versatile synthesis of orthogonally protected derivatives of carba-α-D-glucosamine, carba-α-D-mannose, carba-α-D-mannuronic acid, carba-β-L-idosamine, and carba-β-L-gulose from methyl α-D-mannoside is described. Our synthetic strategy utilizes the palladium-promoted Ferrier carbocyclization and persistent butane-2,3-diacetal protection to produce a key chiral cyclohexanone intermediate, from
ACS applied materials & interfaces, 4(3), 1560-1564 (2012-02-18)
Self-assembled TiO(2) nanoparticulate materials with well-defined spherical morphologies were synthesized by using a biopolymer sodium alginate as a template under different synthesis conditions. Powder X-ray diffraction (XRD), transmission electron microscopy (TEM), and scanning electron microscopy (SEM) techniques were used to
The Journal of organic chemistry, 66(19), 6244-6251 (2001-09-18)
High-resolution (13)C NMR spectra (150 MHz) have been obtained on the complete series of D-aldohexoses (D-allose 1, D-altrose 2, D-galactose 3, D-glucose 4, D-gulose 5, D-idose 6, D-mannose 7, D-talose 8) selectively labeled with (13)C at C1 in order to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service