Skip to Content
Merck
All Photos(1)

Key Documents

00582

Sigma-Aldrich

Ethane

≥99.95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH3
CAS Number:
Molecular Weight:
30.07
Beilstein:
1730716
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

vapor density

1.05 (vs air)

Quality Level

vapor pressure

37.95 atm ( 21.1 °C)

Assay

≥99.95% (GC)

autoignition temp.

881 °F

expl. lim.

13 %

impurities

CnHm, trace
CH4, trace
CO2, trace
H2, trace
N2, trace
O2, trace
water, trace
≤0.04% ethylene

bp

−88 °C (lit.)

mp

−172 °C (lit.)

density

0.362 g/mL at 20 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 l

Other Notes

Sales restrictions may apply

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-211.0 °F - closed cup

Flash Point(C)

-135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Dario Frascari et al.
    Bioresource technology, 128, 479-486 (2012-12-04)
    A novel aerobic/anaerobic/aerobic treatment was implemented in batch reactors containing aquifer materials from a site contaminated by tetrachloroethylene (PCE), trichloroethylene (TCE), vinyl chloride (VC), 1,1,2-trichloroethane (1,1,2-TCA) and chloroform (CF). Consortia grown aerobically on methane, propane, n-pentane and n-hexane completely biodegraded
    Margaret A Greene et al.
    Organic letters, 14(16), 4293-4296 (2012-05-10)
    Stereospecific nickel-catalyzed cross-coupling reactions of benzylic 2-methoxyethyl ethers are reported for the preparation of enantioenriched 1,1-diarylethanes. The 2-methoxyethyl ether serves as a traceless directing group that accelerates cross-coupling. Chelation of magnesium ions is proposed to activate the benzylic C-O bond
    Isobel J Simpson et al.
    Nature, 488(7412), 490-494 (2012-08-24)
    After methane, ethane is the most abundant hydrocarbon in the remote atmosphere. It is a precursor to tropospheric ozone and it influences the atmosphere's oxidative capacity through its reaction with the hydroxyl radical, ethane's primary atmospheric sink. Here we present
    Sachiko Tojo et al.
    The Journal of organic chemistry, 78(5), 1887-1893 (2012-11-29)
    A central carbon–carbon (C–C) σ bond dissociation of polyphenylethane radical anions (Ph(n)E•-, n = 3–6), mesolysis, was investigated by the transient absorption measurement during pulse radiolysis of Ph(n)E in 2-methyltetrahydrofuran. The charge resonance (CR) band of 1,1,2,2-tetraphenylethane radical anion (1,1,2,2-Ph4E•-)
    Prantik Maity et al.
    Journal of the American Chemical Society, 135(1), 280-285 (2012-12-28)
    We have developed a nickel-catalyzed cross coupling of benzylic ammonium triflates with aryl boronic acids to afford diarylmethanes and diarylethanes. This reaction proceeds under mild reaction conditions and with exceptional functional group tolerance. Further, it transforms branched benzylic ammonium salts

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service